Assay Details
Gene Symbol: Bcas3os2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046194.1 | 1 - 2 | 145 | 68 | ||
GenBank mRNA | AK020598.1 | - | 1 - 2 | 146 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 5110 | Mir5110 | mmu-mir-5110 | -6.9kb | Sense | Chr. 11 - 85760616 - 85760702 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 2610027K06 gene | 2610027K06Rik | 16.0kb | Sense | Chr. 11 - 85791660 - 85832388 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
