Assay Details
| Gene Symbol: Msl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015496.1 | - | 1 - 2 | 105 | 59 | |
| AK161412.1 | - | 1 - 2 | 84 | 59 | ||
| DQ387855.1 | - | 1 - 2 | 104 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK161412.1 transcripts.
Set Membership: |
|

Msl1
Genomic Map