Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500018405.mm.1 |
NM_010127.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018405.mm.1 |
NM_010127.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018405.mm.1 |
TC1500002243.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018401.mm.1 |
NM_010127.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018401.mm.1 |
NM_010127.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018401.mm.1 |
TC1500002243.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018404.mm.1 |
NM_010127.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018404.mm.1 |
NM_010127.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018404.mm.1 |
TC1500002243.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018406.mm.1 |
NM_010127.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018406.mm.1 |
NM_010127.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018406.mm.1 |
TC1500002243.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018396.mm.1 |
NM_010127.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018396.mm.1 |
NM_010127.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018396.mm.1 |
TC1500002243.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018399.mm.1 |
NM_010127.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018399.mm.1 |
NM_010127.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018399.mm.1 |
TC1500002243.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018403.mm.1 |
NM_010127.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018403.mm.1 |
NM_010127.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018403.mm.1 |
TC1500002243.mm.1 |
MTA TC |