Assay Details
| Gene Symbol: Tmprss12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183109.3 | NP_898932.2 | 4 - 5 | 809 | 69 | |
| GenBank mRNA | AK132967.1 | - | 4 - 5 | 810 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30484945 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30484946 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30484944 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1997110 | AK015583.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987321 | NM_183109.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987321 | AK132967.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 840033 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 599553 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 194613 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 957005 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712520 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29963 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 619939 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 78472 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164859 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152322 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1997110 | AK015583.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987321 | NM_183109.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987321 | AK132967.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30484956 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08481125 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152322 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164859 | - | - |
| Whole Mouse Genome G4122A | A_51_P439531 | NM_183109.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P439531 | AK015583.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P439531 | AK132967.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135347 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164859 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152322 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12187451 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12131903 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12131901 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30484944 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30484945 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30484946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30484943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12131905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12164818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11994514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11997233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11998201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12087901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11997231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12164814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12151560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12020713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12131908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12164817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12454451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12020708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11909006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30484938 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10426884 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426881 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426887 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426886 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426888 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426883 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426882 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10426880 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500008236.mm.1 | NM_183109.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008236.mm.1 | NM_183109.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008236.mm.1 | TC1500001039.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 169602_f_at | - | - |
| Murine Genome U74v2 Set | 169602_PM_f_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 724604 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4570138 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004570138 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2714612 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2714612 | - | - |
| mouse-6 v1.1 (Illumina) | scl47508.3.1_182-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4570138 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47508.3.1_182-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4570138 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2797098 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2714612 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2797098 | - | - |

Tmprss12
Genomic Map