Assay Details
| Gene Symbol: Pacsin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286743.1 | NP_001273672.1 | 6 - 7 | 1107 | 71 | |
| NM_001286744.1 | NP_001273673.1 | 6 - 7 | 1013 | 71 | ||
| NM_011861.3 | NP_035991.1 | 6 - 7 | 1072 | 71 | ||
| NM_178365.4 | NP_848142.1 | 6 - 7 | 1090 | 71 | ||
| GenBank mRNA | AK043967.1 | - | 6 - 7 | 1076 | 71 | |
| AK077958.1 | - | 1 - 2 | 192 | 71 | ||
| AK158686.1 | - | 6 - 7 | 1110 | 71 | ||
| BC014698.1 | - | 6 - 7 | 961 | 4527708 | 71 | |
| X85124.1 | - | 6 - 7 | 981 | 71 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pacsin1
Genomic Map