Assay Details
Gene Symbol: Pim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008842.3 | NP_032868.2 | 2 - 3 | 289 | 75 | |
GenBank mRNA | AK154669.1 | - | 1 - 2 | 128 | 75 | |
AK158445.1 | - | 2 - 3 | 541 | 75 | ||
BC042885.1 | - | 2 - 3 | 522 | 4507947 | 75 | |
BC053019.1 | - | 2 - 3 | 289 | 5694941 | 75 | |
BC055316.1 | - | 2 - 3 | 289 | 5694941 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
