Assay Details
Gene Symbol: Rab44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002786.2 | NP_001002786.1 | 4 - 5 | 3019 | 70 | |
GenBank mRNA | AK036563.1 | - | 4 - 5 | 3020 | 70 | |
BC139372.1 | - | 4 - 5 | 2627 | 8862394 | 70 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
