Assay Details
Gene Symbol: Rab44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002786.2 | NP_001002786.1 | 6 - 7 | 3236 | 60 | |
GenBank mRNA | AK036563.1 | - | 6 - 7 | 3237 | 60 | |
BC139372.1 | - | 6 - 7 | 2844 | 8862394 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
