Assay Details
Gene Symbol: Rab44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002786.2 | NP_001002786.1 | 7 - 8 | 3311 | 60 | |
GenBank mRNA | AK036563.1 | - | 7 - 8 | 3312 | 60 | |
BC139372.1 | - | 7 - 8 | 2919 | 8862394 | 60 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000409 | NM_001002786.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000409 | AK036563.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 950464 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 512456 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741060 | AK081816.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741060 | BC010758.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741060 | D26123.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741060 | NM_007621.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741060 | X07411.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 269499 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558274 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579105 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 599537 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904473 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227264 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 44565 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177554 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143179 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2000409 | NM_001002786.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2000409 | AK036563.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177554 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143179 | - | - |
Whole Mouse Genome G4122A | A_51_P493816 | NM_001002786.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P493816 | AK036563.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P493816 | BC139372.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122338 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177554 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126123 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126166 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126173 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31126193 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126121 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126232 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126169 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13457709 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126185 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126145 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126229 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126139 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126246 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126177 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126236 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13378744 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126137 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126173 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13426244 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126159 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13345153 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13403591 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126128 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126180 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126188 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126223 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13071477 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126252 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126244 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13559615 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126233 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126127 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126183 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126194 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13364901 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126136 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126134 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126192 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126144 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126168 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13432958 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126171 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126175 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13345151 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126228 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126154 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126257 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126218 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126220 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126258 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13241753 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126227 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126123 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126184 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126130 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13376484 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13571155 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126166 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13386805 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126156 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126226 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126118 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126146 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126131 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126151 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126164 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126249 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126174 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13426245 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31126217 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13364898 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13378753 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10443477 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10443473 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10443478 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10443475 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10443474 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10443472 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10443476 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10443471 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1456980_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10443470 | - | - |
Mouse Genome 430 2.0 Array | 1456980_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1456980_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 442827_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 442827_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700004393.mm.1 | NM_001002786.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700004393.mm.1 | NM_001002786.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700004393.mm.1 | TC1700000498.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700004394.mm.1 | NM_001002786.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700004394.mm.1 | NM_001002786.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700004394.mm.1 | TC1700000498.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700004395.mm.1 | NM_001002786.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700004395.mm.1 | NM_001002786.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700004395.mm.1 | TC1700000498.mm.1 | MTA TC |
Mouse Expression Set 430 | 1456980_PM_at | - | - |
Mouse Expression Set 430 | 1456980_at | - | - |
Murine Genome U74v2 Set | 129148_at | - | - |
Murine Genome U74v2 Set | 129148_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456980_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456980_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 369798 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 102230136 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 102230136 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1216928 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102230136 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102230136 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3076909 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3155815 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3155815 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3076909 | - | - |
