Assay Details
Gene Symbol: Taf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026836.2 | NP_081112.1 | 2 - 3 | 373 | 73 | |
GenBank mRNA | AK050689.1 | - | 2 - 3 | 374 | 73 | |
AK133851.1 | - | 3 - 4 | 1357 | 73 | ||
BC005603.1 | - | 2 - 3 | 352 | 3483315 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
