Assay Details
Gene Symbol: Wbp2nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029066.1 | NP_083342.1 | 1 - 2 | 77 | 94 | |
GenBank mRNA | AK015863.1 | - | 1 - 2 | 108 | 94 | |
BC119520.2 | - | 1 - 2 | 77 | 40055925 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
