Assay Details
Gene Symbol: Itga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033228.3 | NP_001028400.2 | 18 - 19 | 2825 | 66 | |
GenBank mRNA | AK035870.1 | - | 18 - 19 | 2826 | 66 | |
AK040830.1 | - | 9 - 10 | 1249 | 66 | ||
AK137169.1 | - | 18 - 19 | 2590 | 66 | ||
DQ113653.1 | - | 1 - 2 | 82 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
