Assay Details
| Gene Symbol: Mettl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144877.1 | NP_659126.1 | 2 - 3 | 994 | 81 | |
| GenBank mRNA | AK050492.1 | - | 2 - 3 | 1187 | 81 | |
| AK146751.1 | - | 1192 | 81 | |||
| AK162843.1 | - | 2 - 3 | 1179 | 81 | ||
| AK168959.1 | - | 2 - 3 | 1201 | 81 | ||
| AK169319.1 | - | 1193 | 81 | |||
| AK173044.1 | - | 1147 | 81 | |||
| BC014872.1 | - | 2 - 3 | 994 | 3602469 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl13
Genomic Map