Assay Details
| Gene Symbol: Mettl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144877.1 | NP_659126.1 | 6 - 7 | 1779 | 97 | |
| GenBank mRNA | AK084750.1 | - | 6 - 7 | 1474 | 97 | |
| AK146751.1 | - | 1977 | 97 | |||
| AK168959.1 | - | 6 - 7 | 1986 | 97 | ||
| AK169319.1 | - | 1978 | 97 | |||
| AK173044.1 | - | 1932 | 97 | |||
| BC014872.1 | - | 6 - 7 | 1779 | 3602469 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl13
Genomic Map