Assay Details
Gene Symbol: Gpbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122963.1 | NP_001116435.1 | 9 - 10 | 2308 | 141 | |
NM_028487.4 | NP_082763.3 | 10 - 11 | 2368 | 141 | ||
GenBank mRNA | AK135728.1 | - | 10 - 11 | 2299 | 141 | |
AK136894.1 | - | 10 - 11 | 2148 | 141 | ||
AK145318.1 | - | 9 - 10 | 2297 | 141 | ||
AK151283.1 | - | 8 - 9 | 1529 | 141 | ||
AK163370.1 | - | 10 - 11 | 2216 | 141 | ||
AK163390.1 | - | 10 - 11 | 2370 | 141 | ||
AK169406.1 | - | 10 - 11 | 2163 | 141 | ||
AY382529.1 | - | 10 - 11 | 2166 | 141 | ||
BC016083.1 | - | 8 - 9 | 1162 | 4501399 | 141 | |
BC067075.1 | - | 9 - 10 | 2082 | 30614988 | 141 | |
BC076635.1 | - | 9 - 10 | 2138 | 141 | ||
BC094888.1 | - | 9 - 10 | 2101 | 30628044 | 141 | |
BC096442.1 | - | 10 - 11 | 2183 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
