Assay Details
Gene Symbol: Gpbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122963.1 | NP_001116435.1 | 1 - 2 | 326 | 149 | |
NM_028487.4 | NP_082763.3 | 1 - 2 | 326 | 149 | ||
GenBank mRNA | AK006608.1 | - | 1 - 2 | 343 | 149 | |
AK082704.1 | - | 1 - 2 | 144 | 149 | ||
AK135728.1 | - | 1 - 2 | 255 | 149 | ||
AK136894.1 | - | 1 - 2 | 104 | 150 | ||
AK145318.1 | - | 1 - 2 | 314 | 149 | ||
AK161928.1 | - | 1 - 2 | 113 | 149 | ||
AK163370.1 | - | 1 - 2 | 174 | 149 | ||
AK163390.1 | - | 1 - 2 | 326 | 149 | ||
AK169406.1 | - | 1 - 2 | 121 | 149 | ||
AY382529.1 | - | 1 - 2 | 124 | 149 | ||
BC067075.1 | - | 1 - 2 | 100 | 30614988 | 149 | |
BC076635.1 | - | 1 - 2 | 155 | 149 | ||
BC094888.1 | - | 1 - 2 | 119 | 30628044 | 149 | |
BC096442.1 | - | 1 - 2 | 143 | 149 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK082704.1, AK135728.1, AK145318.1, AK161928.1, AK163370.1, AK163390.1, AK169406.1, AY382529.1, BC067075.1, BC094888.1, NM_001122963.1, NM_028487.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
