Assay Details
| Gene Symbol: Krt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008470.1 | NP_032496.1 | 6 - 7 | 1283 | 96 | |
| GenBank mRNA | AF053235.1 | - | 6 - 7 | 1283 | 96 | |
| BC103666.1 | - | 6 - 7 | 1283 | 40046212 | 96 | |
| BC114977.1 | - | 6 - 7 | 1283 | 40046220 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krt16
Genomic Map