Assay Details
| Gene Symbol: Htr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008308.4 | NP_032334.2 | 1 | 2803 | 86 | |
| GenBank mRNA | AK034017.1 | - | 1 | 2251 | 86 | |
| AK043668.1 | - | 1 | 2262 | 86 | ||
| AK049814.1 | - | 1 | 2643 | 86 | ||
| AK049884.1 | - | 1 | 2734 | 86 | ||
| AK140194.1 | - | 1 | 2857 | 86 | ||
| BC138669.1 | - | 1 | 2510 | 8861690 | 86 | |
| BC138681.1 | - | 1 | 2510 | 8861702 | 86 | |
| U39391.1 | - | 1 | 2312 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Htr1a
Genomic Map