Assay Details
| Gene Symbol: Ikzf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011771.1 | NP_035901.1 | 6 - 7 | 989 | 83 | |
| GenBank mRNA | AF001293.1 | - | 5 - 6 | 708 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ikzf3
Genomic Map