Assay Details
| Gene Symbol: Grb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010346.2 | NP_034476.1 | 5 - 6 | 1007 | 94 | |
| GenBank mRNA | AK145996.1 | - | 5 - 6 | 823 | 94 | |
| AK168438.1 | - | 5 - 6 | 791 | 94 | ||
| BC003295.1 | - | 5 - 6 | 750 | 3583896 | 94 | |
| BC081547.1 | - | 5 - 6 | 793 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grb7
Genomic Map