Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100014378.mm.1 |
NM_001109626.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014378.mm.1 |
NM_001109626.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014378.mm.1 |
NM_001109628.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014378.mm.1 |
NM_001109628.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014378.mm.1 |
NM_026952.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014378.mm.1 |
NM_026952.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014378.mm.1 |
TC1100001560.mm.1 |
MTA TC |