Assay Details
Gene Symbol: Erbb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003817.1 | NP_001003817.1 | 16 - 17 | 2123 | 73 | |
GenBank mRNA | AK031099.1 | - | 16 - 17 | 1991 | 73 | |
AK031542.1 | - | 8 - 9 | 821 | 73 | ||
AK083669.1 | - | 16 - 17 | 1992 | 73 | ||
AK129487.1 | - | 16 - 17 | 2093 | 73 | ||
AK142296.1 | - | 16 - 17 | 2117 | 74 | ||
BC046553.1 | - | 2 - 3 | 478 | 73 | ||
BC046811.1 | - | 16 - 17 | 2114 | 5702040 | 73 | |
BC053078.1 | - | 16 - 17 | 2114 | 5702040 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
