Assay Details
Gene Symbol: Erbb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003817.1 | NP_001003817.1 | 18 - 19 | 2393 | 60 | |
GenBank mRNA | AK031099.1 | - | 18 - 19 | 2261 | 60 | |
AK031542.1 | - | 10 - 11 | 1091 | 60 | ||
AK083669.1 | - | 17 - 18 | 2343 | 60 | ||
AK129487.1 | - | 18 - 19 | 2363 | 60 | ||
AK142296.1 | - | 18 - 19 | 2387 | 60 | ||
BC046553.1 | - | 4 - 5 | 748 | 60 | ||
BC046811.1 | - | 17 - 18 | 2465 | 5702040 | 60 | |
BC053078.1 | - | 17 - 18 | 2465 | 5702040 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
