Assay Details
| Gene Symbol: Kcnh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081194.2 | NP_001074663.1 | 10 - 11 | 2009 | 62 | |
| GenBank mRNA | BC150990.1 | - | 10 - 11 | 2017 | 9087903 | 62 |
| BC150994.1 | - | 10 - 11 | 2013 | 9087907 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnh4
Genomic Map