Assay Details
Gene Symbol: Kcnh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081194.2 | NP_001074663.1 | 5 - 6 | 1006 | 60 | |
GenBank mRNA | BC150990.1 | - | 5 - 6 | 1014 | 9087903 | 60 |
BC150994.1 | - | 5 - 6 | 1010 | 9087907 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
