Assay Details
| Gene Symbol: Fkbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163481.1 | NP_001156953.1 | 7 - 8 | 1340 | 61 | |
| NM_010221.2 | NP_034351.2 | 7 - 8 | 1343 | 61 | ||
| GenBank mRNA | AK019446.2 | - | 7 - 8 | 1282 | 61 | |
| AK144185.1 | - | 7 - 8 | 1323 | 61 | ||
| BC029546.1 | - | 7 - 8 | 1290 | 5059670 | 61 | |
| L07063.1 | - | 7 - 8 | 1346 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fkbp10
Genomic Map