Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100033066.mm.1 |
NM_172300.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033066.mm.1 |
NM_172300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033066.mm.1 |
TC1100003609.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100033067.mm.1 |
NM_172300.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033067.mm.1 |
NM_172300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033067.mm.1 |
TC1100003609.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100033065.mm.1 |
NM_172300.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033065.mm.1 |
NM_172300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033065.mm.1 |
TC1100003609.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100033064.mm.1 |
NM_172300.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033064.mm.1 |
NM_172300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033064.mm.1 |
TC1100003609.mm.1 |
MTA TC |