Assay Details
Gene Symbol: Prr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175563.5 | NP_780772.1 | 9 - 10 | 1122 | 77 | |
GenBank mRNA | AK040348.1 | - | 9 - 10 | 1100 | 77 | |
AK047463.1 | - | 9 - 10 | 1100 | 77 | ||
AK083651.1 | - | 9 - 10 | 1117 | 77 | ||
BC023918.1 | - | 9 - 10 | 1132 | 5323297 | 77 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992591 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992592 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992594 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992593 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069221 | BC023918.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069221 | NM_175563.5 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 856505 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 755958 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441035 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 806155 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 762168 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552956 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 690648 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 480746 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 517746 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 4158 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10342 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 59835 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115754 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069221 | BC023918.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069221 | NM_175563.5 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992517 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992515 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992592 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992510 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992588 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992583 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992571 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992587 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992512 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992511 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992591 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992570 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992513 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992582 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992507 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992577 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992595 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992567 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992580 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992519 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992576 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992586 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992572 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02386376 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07430325 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07430299 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115754 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02386364 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07430308 | - | - |
Whole Mouse Genome G4122A | A_51_P483269 | BC023918.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P483269 | AK047463.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P483269 | AK083651.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P483269 | NM_175563.5 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P483269 | AK040348.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P338618 | BC023918.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P338618 | AK047463.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P338618 | AK083651.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P338618 | NM_175563.5 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P338618 | AK040348.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P33097 | BC023918.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P33097 | AK083651.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P33097 | NM_175563.5 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132313 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103502 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115754 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02386376 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09481065 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09481057 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09481074 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09481062 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992591 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992594 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992592 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992578 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992553 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992589 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992551 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09613180 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09480515 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08986710 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992519 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992529 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992553 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992531 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992524 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09481074 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992515 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992557 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09063350 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09324131 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09512747 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992522 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992555 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992564 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992558 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992589 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992562 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09584476 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992510 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992576 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992586 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992568 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992517 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992584 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992514 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992570 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992578 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992552 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992507 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09512749 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992513 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09718399 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992549 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992580 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992511 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09512750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992583 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27992588 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10389610 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10389616 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10389615 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10389614 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10389613 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10389607 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10389612 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10389611 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10389609 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10389608 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1444257_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1442134_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10389606 | - | - |
Mouse Genome 430 2.0 Array | 1444257_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1444257_at | - | - |
Mouse Genome 430 2.0 Array | 1442134_at | - | - |
Mouse Genome 430 2.0 Array | 1442134_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 270906_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 270906_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100031860.mm.1 | NM_175563.5.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100031860.mm.1 | NM_175563.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100031860.mm.1 | TC1100003470.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100031859.mm.1 | NM_175563.5.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100031859.mm.1 | NM_175563.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100031859.mm.1 | TC1100003470.mm.1 | MTA TC |
Mouse Expression Set 430 | 1442134_PM_at | - | - |
Mouse Expression Set 430 | 1444257_at | - | - |
Mouse Expression Set 430 | 1444257_PM_at | - | - |
Mouse Expression Set 430 | 1442134_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1442134_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1442134_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1444257_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1444257_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 480841 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1340373 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1340373 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001340373 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225967 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1225967 | - | - |
mouse-6 v1.1 (Illumina) | scl39765.10_275-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1340373 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl39765.10_275-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1340373 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2926068 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2926068 | - | - |
