Assay Details
Gene Symbol: Dhx40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026191.2 | NP_080467.3 | 3 - 4 | 533 | 150 | |
GenBank mRNA | AK004610.1 | - | 3 - 4 | 497 | 150 | |
AK010512.1 | - | 3 - 4 | 513 | 150 | ||
AK029935.1 | - | 3 - 4 | 506 | 150 | ||
AK031000.1 | - | 3 - 4 | 480 | 150 | ||
AK135592.1 | - | 3 - 4 | 469 | 150 | ||
BC058276.1 | - | 3 - 4 | 451 | 6515479 | 150 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK031000.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
