Assay Details
| Gene Symbol: Cyb5r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029787.2 | NP_084063.1 | 3 - 4 | 302 | 75 | |
| GenBank mRNA | AF332059.1 | - | 3 - 4 | 226 | 75 | |
| AF332060.1 | - | 3 - 4 | 226 | 75 | ||
| AK002640.1 | - | 3 - 4 | 274 | 75 | ||
| AK010858.1 | - | 3 - 4 | 279 | 75 | ||
| AK128917.1 | - | 3 - 4 | 296 | 75 | ||
| BC004760.1 | - | 3 - 4 | 309 | 3154531 | 75 | |
| BC032013.1 | - | 3 - 4 | 284 | 4238275 | 75 | |
| BC043074.1 | - | 3 - 4 | 302 | 6404746 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyb5r3
Genomic Map