Assay Details
| Gene Symbol: A4galt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004150.3 | NP_001004150.1 | 2 - 3 | 217 | 66 | |
| NM_001170954.1 | NP_001164425.1 | 2 - 3 | 257 | 66 | ||
| GenBank mRNA | AK139833.1 | - | 2 - 3 | 220 | 66 | |
| AK149186.1 | - | 2 - 3 | 173 | 66 | ||
| AK162529.1 | - | 2 - 3 | 256 | 66 | ||
| AY371179.1 | - | 2 - 3 | 176 | 66 | ||
| BC138844.1 | - | 1 - 2 | 132 | 8861866 | 66 | |
| BC138845.1 | - | 1 - 2 | 132 | 8861867 | 66 | |
| BY770781.1 | - | 2 - 3 | 259 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK149186.1, AK162529.1, AY371179.1, BC138844.1, BC138845.1, NM_001004150.3, NM_001170954.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30387346 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30387339 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30387340 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30387341 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30387343 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30387338 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30387345 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P130310 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773256 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 560104 | AK002880.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 560104 | AK028131.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 560104 | AK077235.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 560104 | BC003795.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 560104 | L41154.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 560104 | NM_011526.4 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 560104 | U36588.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 153319 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609398 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 16684 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 707759 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 758234 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782409 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352471 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782619 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529555 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 289683 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587266 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125080 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 148072 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115083 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P130310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30387340 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03085099 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115083 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08439758 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08439737 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115083 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12382333 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12382329 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12130018 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12130020 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30387338 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30387346 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30387345 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30387343 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30387341 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30387340 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30387339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30387206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12326566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12088085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11966696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12134237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387386 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12382323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387347 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12447493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12130018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12318543 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12355981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12050064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12134235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12447499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12382332 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12318533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12311760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12407321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12318541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11967773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12088083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387316 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12382342 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30387388 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10430969 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430973 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430970 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430972 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430971 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455338_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10430968 | - | - |
| Mouse Genome 430 2.0 Array | 1455338_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455338_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 239559_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 239559_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500015529.mm.1 | NM_001004150.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015529.mm.1 | NM_001004150.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015529.mm.1 | NM_001170954.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015529.mm.1 | NM_001170954.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015529.mm.1 | TC1500001968.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015528.mm.1 | NM_001004150.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015528.mm.1 | NM_001004150.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015528.mm.1 | NM_001170954.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015528.mm.1 | NM_001170954.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015528.mm.1 | TC1500001968.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015534.mm.1 | NM_001004150.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015534.mm.1 | NM_001004150.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015534.mm.1 | TC1500001968.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455338_PM_at | - | - |
| Mouse Expression Set 430 | 1455338_at | - | - |
| Murine Genome U74v2 Set | 161085_r_at | - | - |
| Murine Genome U74v2 Set | 161085_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455338_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455338_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455338_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455338_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 356220 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1218739 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3009373 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3009373 | - | - |

A4galt
Genomic Map