Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500000246.mm.1 |
NM_001008702.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000246.mm.1 |
NM_001008702.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000246.mm.1 |
NM_001037905.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000246.mm.1 |
NM_001037905.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000246.mm.1 |
NM_023118.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000246.mm.1 |
NM_023118.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000246.mm.1 |
TC1500000031.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500000245.mm.1 |
NM_001008702.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000245.mm.1 |
NM_001008702.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000245.mm.1 |
NM_001037905.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000245.mm.1 |
NM_001037905.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000245.mm.1 |
NM_001102400.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000245.mm.1 |
NM_001102400.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000245.mm.1 |
NM_023118.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000245.mm.1 |
NM_023118.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000245.mm.1 |
TC1500000031.mm.1 |
MTA TC |