Assay Details
Gene Symbol: 4921509O07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045501.1 | 5 - 6 | 1706 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cDNA sequence BC067074 | BC067074 | -80.4kb | Anti-sense | Chr. 13 - 113292430 - 113370515 on NCBI Build GRCm38 | |
Downstream |
predicted gene, 34471 |
Gm34471 | 42.8kb | Sense | Chr. 13 - 113507828 - 113568698 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
