Assay Details
| Gene Symbol: Zfp641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173769.3 | NP_776130.1 | 1 - 2 | 26 | 58 | |
| GenBank mRNA | AK036826.1 | - | 1 - 2 | 27 | 58 | |
| AK087530.1 | - | 1 - 2 | 74 | 58 | ||
| AK136402.1 | - | 1 - 2 | 104 | 58 | ||
| BC111896.1 | - | 1 - 2 | 26 | 40061302 | 58 | |
| BC112418.1 | - | 1 - 2 | 26 | 40061301 | 58 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK036826.1, AK136402.1, BC111896.1, BC112418.1, NM_173769.3 transcripts.
Set Membership: |
|

Zfp641
Genomic Map