Assay Details
| Gene Symbol: Zfp641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173769.3 | NP_776130.1 | 4 - 5 | 460 | 73 | |
| GenBank mRNA | AK036826.1 | - | 4 - 5 | 461 | 73 | |
| AK087530.1 | - | 4 - 5 | 508 | 73 | ||
| AK136402.1 | - | 4 - 5 | 538 | 73 | ||
| BC111896.1 | - | 4 - 5 | 418 | 40061302 | 73 | |
| BC112418.1 | - | 4 - 5 | 418 | 40061301 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
RNA polymerase II transcription factor activity, sequence-specific DNA binding
nucleic acid binding
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
RNA polymerase II transcription factor activity, sequence-specific DNA binding
nucleic acid binding
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30474028 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30474023 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30474027 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30474025 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30474024 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30474026 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P392644 | AK036826.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P392644 | AK087530.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P392644 | NM_173769.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P392644 | AK036826.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P392644 | AK087530.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P392644 | NM_173769.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 4454 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364668 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396076 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 86737 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 407215 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 8716 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 67442 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140745 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146489 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P392644 | AK036826.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P392644 | AK087530.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P392644 | NM_173769.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P392644 | AK036826.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P392644 | AK087530.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P392644 | NM_173769.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474017 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146489 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140745 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08475965 | - | - |
| Whole Mouse Genome G4122A | A_52_P392644 | AK036826.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P392644 | AK087530.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P392644 | NM_173769.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P392644 | AK036826.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P392644 | AK087530.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P392644 | NM_173769.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134231 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112328 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140745 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146489 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12444092 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12444104 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12444095 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12013002 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12444105 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30474025 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30474028 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30474027 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30474023 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30474024 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30474026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473985 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12450031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12309463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12275127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12463132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12309469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11849820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12309456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11938920 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10432141 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432148 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432143 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432146 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432145 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432147 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432144 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432142 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10432139 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500017619.mm.1 | NM_173769.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500017619.mm.1 | NM_173769.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500017619.mm.1 | TC1500002170.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500017617.mm.1 | NM_173769.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500017617.mm.1 | NM_173769.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500017617.mm.1 | TC1500002170.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500017618.mm.1 | NM_173769.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500017618.mm.1 | NM_173769.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500017618.mm.1 | TC1500002170.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 381063 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 103990497 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4060338 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004060338 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1256695 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2493040 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1256695 | - | - |
| mouse-6 v1.1 (Illumina) | scl00239652.2_53-S | - | - |
| mouse-6 v1.1 (Illumina) | scl46772.7_573-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4060338 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103990497 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00239652.2_53-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4060338 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103990497 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1256695 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1256695 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00239652.2_53-S | - | - |

Zfp641
Genomic Map