Assay Details
| Gene Symbol: Ptrh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK038888.1 | - | 1 - 2 | 90 | 104 | |
 Back To Top
 Back To TopMore Information
Additional Information:
				    	 
					    	
					        	
					            	This assay is in the 5'UTR region of the AK038888.1 transcripts.
					        	
					    	            
				    	
			    	
			    
			| Set Membership: |  Amplicon length greater than or equal to 101  3' Most  Mus musculus  Probe spans exons | 
 Back To Top
 Back To Top
 Ptrh2
Ptrh2 Genomic Map
Genomic Map Back To Top
 Back To Top