Assay Details
| Gene Symbol: Lmf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178919.4 | NP_849250.1 | 3 - 4 | 590 | 113 | |
| GenBank mRNA | AK042427.1 | - | 3 - 4 | 549 | 114 | |
| AK084606.1 | - | 3 - 4 | 416 | 113 | ||
| AK085435.1 | - | 3 - 4 | 541 | 114 | ||
| AK149916.1 | - | 3 - 4 | 542 | 113 | ||
| AK149934.1 | - | 3 - 4 | 591 | 113 | ||
| AK171488.1 | - | 3 - 4 | 571 | 113 | ||
| BC038039.1 | - | 2 - 3 | 1092 | 113 | ||
| BC058973.1 | - | 1 - 2 | 724 | 113 | ||
| BC079532.1 | - | 3 - 4 | 486 | 6403184 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmf2
Genomic Map