Assay Details
| Gene Symbol: Lmf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178919.4 | NP_849250.1 | 8 - 9 | 1358 | 61 | |
| GenBank mRNA | AK042427.1 | - | 8 - 9 | 1318 | 61 | |
| AK084606.1 | - | 8 - 9 | 1184 | 61 | ||
| AK085435.1 | - | 8 - 9 | 1310 | 61 | ||
| AK149916.1 | - | 8 - 9 | 1310 | 61 | ||
| AK149934.1 | - | 8 - 9 | 1359 | 61 | ||
| AK158371.1 | - | 7 - 8 | 1413 | 61 | ||
| AK171488.1 | - | 8 - 9 | 1339 | 61 | ||
| BC026448.1 | - | 7 - 8 | 1138 | 61 | ||
| BC038039.1 | - | 7 - 8 | 1860 | 61 | ||
| BC058973.1 | - | 6 - 7 | 1492 | 61 | ||
| BC079532.1 | - | 8 - 9 | 1254 | 6403184 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmf2
Genomic Map