Assay Details
Gene Symbol: Selo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027905.2 | NP_082181.2 | 2 - 3 | 767 | 69 | |
GenBank mRNA | AK005048.1 | - | 2 - 3 | 767 | 69 | |
BC117817.1 | - | 3 - 4 | 282 | 69 | ||
BC117818.1 | - | 3 - 4 | 315 | 69 | ||
BC141293.1 | - | 2 - 3 | 836 | 9055905 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
