Assay Details
Gene Symbol: Mum1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164630.1 | NP_001158102.1 | 7 | 3269 | 135 | |
NM_001164631.1 | NP_001158103.1 | 4 | 2851 | 135 | ||
NM_001164632.1 | NP_001158104.1 | 5 | 3012 | 135 | ||
NM_001164633.1 | NP_001158105.1 | 5 | 2951 | 135 | ||
NM_175541.3 | NP_780750.2 | 6 | 3192 | 135 | ||
GenBank mRNA | AK035086.1 | - | 1 | 2020 | 135 | |
BC053077.1 | - | 1 | 1159 | 135 | ||
BC063766.1 | - | 1 | 1462 | 135 | ||
BC068178.1 | - | 4 | 2870 | 135 | ||
BC096534.1 | - | 1 | 2194 | 135 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020090 | BC096534.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020090 | BC053077.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020090 | BC063766.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020090 | BC068178.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020090 | AK035086.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 134639 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385217 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133439 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020090 | BC096534.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020090 | BC053077.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020090 | BC063766.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020090 | BC068178.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020090 | AK035086.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133439 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09435504 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09435496 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09435479 | - | - |
Whole Mouse Genome G4122A | A_51_P418029 | BC096534.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P418029 | BC053077.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P418029 | BC063766.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P418029 | BC068178.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P418029 | AK035086.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135023 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09435496 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693317 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693338 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14856010 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693174 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693214 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693294 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693231 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693157 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693171 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693222 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693228 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693202 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693154 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693200 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693172 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693177 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693144 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693225 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693184 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693151 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14855994 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14876252 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14815214 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693159 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693205 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14565653 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693297 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693220 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693250 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693298 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693163 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693198 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693148 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693156 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693247 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693187 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693208 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693244 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693188 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693217 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693289 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693302 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693242 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693215 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693296 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693204 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693149 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693167 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15169412 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693175 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693166 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693179 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693211 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693178 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693161 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693226 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693153 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693292 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693191 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693212 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693201 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693304 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14573426 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693224 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693216 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693229 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693235 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693306 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693210 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693195 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693183 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693181 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14855989 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693142 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693299 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14835044 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693305 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15169414 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14876253 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693223 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693237 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693301 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693162 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693241 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693219 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693169 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693288 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10601986 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10601985 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10601984 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10601983 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10601987 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1455238_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10601980 | - | - |
Mouse Genome 430 2.0 Array | 1455238_at | - | - |
Mouse Genome 430 2.0 Array | 1455238_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 245631_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 245631_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0X00008598.mm.1 | NM_001164630.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00008598.mm.1 | NM_001164630.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00008598.mm.1 | NM_001164631.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00008598.mm.1 | NM_001164631.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00008598.mm.1 | NM_001164632.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00008598.mm.1 | NM_001164632.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00008598.mm.1 | NM_001164633.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00008598.mm.1 | NM_001164633.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00008598.mm.1 | NM_175541.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00008598.mm.1 | NM_175541.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00008598.mm.1 | TC0X00001367.mm.1 | MTA TC |
Mouse Expression Set 430 | 1455238_PM_at | - | - |
Mouse Expression Set 430 | 1455238_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455238_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455238_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 100540154 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4010411 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004010411 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2604603 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1253308 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2604603 | - | - |
mouse-6 v1.1 (Illumina) | scl0245631.2_148-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4010411 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 100540154 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0245631.2_148-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4010411 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100540154 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2604603 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2604603 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1253308 | - | - |
