Assay Details
Gene Symbol: Trabd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026485.2 | NP_080761.2 | 2 - 3 | 149 | 68 | |
GenBank mRNA | AK005300.1 | - | 2 - 3 | 119 | 68 | |
AK017745.1 | - | 2 - 3 | 143 | 68 | ||
AK146027.1 | - | 2 - 3 | 143 | 68 | ||
BC005574.1 | - | 2 - 3 | 175 | 3967961 | 68 | |
BU510197.1 | - | 2 - 3 | 229 | 68 | ||
CX200611.1 | - | 2 - 3 | 134 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK005300.1, AK017745.1, BC005574.1, NM_026485.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
