Assay Details
Gene Symbol: Trabd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026485.2 | NP_080761.2 | 8 - 9 | 958 | 62 | |
GenBank mRNA | AK131116.1 | - | 1 - 2 | 2922 | 62 | |
AK146027.1 | - | 8 - 9 | 974 | 62 | ||
BC005574.1 | - | 8 - 9 | 984 | 3967961 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
