Assay Details
| Gene Symbol: Syt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018803.2 | NP_061273.1 | 5 - 6 | 1465 | 66 | |
| GenBank mRNA | AB026807.1 | - | 5 - 6 | 1374 | 66 | |
| BC125632.1 | - | 5 - 6 | 1407 | 40130147 | 66 | |
| BC125634.1 | - | 5 - 6 | 1411 | 40130149 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syt10
Genomic Map