Assay Details
| Gene Symbol: Shank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021423.3 | NP_067398.2 | 4 - 5 | 446 | 78 | |
| GenBank mRNA | AB231013.1 | - | 4 - 5 | 446 | 78 | |
| AJ245904.1 | - | 2 - 3 | 300 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shank3
Genomic Map