Assay Details
| Gene Symbol: Mapk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011161.5 | NP_035291.4 | 3 - 4 | 396 | 64 | |
| GenBank mRNA | AF135185.1 | - | 3 - 4 | 376 | 64 | |
| AK144846.1 | - | 3 - 4 | 369 | 64 | ||
| AK161190.1 | - | 3 - 4 | 284 | 64 | ||
| AK161562.1 | - | 3 - 4 | 348 | 64 | ||
| BC057211.1 | - | 3 - 4 | 332 | 64 | ||
| BC064737.1 | - | 3 - 4 | 347 | 30112690 | 64 | |
| BC092526.1 | - | 3 - 4 | 396 | 6593321 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mapk11
Genomic Map