Assay Details
Gene Symbol: Mcidas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037914.3 | NP_001033003.1 | 2 - 3 | 220 | 79 | |
GenBank mRNA | AK134107.1 | - | 2 - 3 | 221 | 79 | |
BC150962.1 | - | 2 - 3 | 243 | 9087873 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
