Assay Details
Gene Symbol: Mcidas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC150962.1 | - | 6 - 7 | 727 | 9087873 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mcidas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC150962.1 | - | 6 - 7 | 727 | 9087873 | 113 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |