Assay Details
| Gene Symbol: 3830408C21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015471.1 | 2 - 3 | 1468 | 70 | ||
| GenBank mRNA | AK014423.1 | - | 2 - 3 | 1468 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

3830408C21Rik
Genomic Map