Assay Details
| Gene Symbol: Gm13119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034101.2 | NP_001029273.2 | 2 - 3 | 332 | 63 | |
| GenBank mRNA | BC100406.1 | - | 1 - 2 | 332 | 30727972 | 63 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm13119
Genomic Map