Assay Details
Gene Symbol: Ampd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033303.2 | NP_001028475.2 | 9 - 10 | 1303 | 116 | |
GenBank mRNA | AK132524.1 | - | 9 - 10 | 1294 | 116 | |
AK137549.1 | - | 9 - 10 | 1295 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
